Anti-BCL10 Rabbit Polyclonal Antibody
89377-618
: Abgent
- Antibody Type:Primary
- Antigen Name:B-cell lymphoma/leukemia 10
- Antigen Symbol:BCL10
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:c-E10|B-cell lymphoma/leukemia 10|BCL-10|mE10|B-cell CLL/lymphoma 10|CIPER|CLAP|CARMEN
- Cat. No.:89377-618
- Supplier no.:AP10698A
Type: Primary
Antigen: BCL10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse