Anti-SREBF2 Rabbit Polyclonal Antibody
Catalog # 89377-560
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Sterol Regulatory Element Binding Transcription Factor 2
- Antigen Symbol:SREBF2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:Class D basic helix-loop-helix protein 2|BHLHD2|bHLHd2|SREBP-2|SREBP2
- Cat. No.:89377-560
- Supplier no.:AP10673C
Specifications
About this item
Type: Primary
Antigen: SREBF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human