Anti-DCAF10 Rabbit Polyclonal Antibody
89377-474
: Abgent
- Antibody Type:Primary
- Antigen Name:DDB1 And CUL4 associated factor 10
- Antigen Symbol:DCAF10
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:WDR32
- Cat. No.:89377-474
- Supplier no.:AP10636B
Type: Primary
Antigen: DCAF10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human