Anti-HFM1 Rabbit Polyclonal Antibody
89377-426
: Abgent
- Antibody Type:Primary
- Antigen Name:HFM1
- Antigen Symbol:HFM1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-426
- Supplier no.:AP10614B
Type: Primary
Antigen: HFM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human