Anti-CCDC107 Rabbit Polyclonal Antibody
Catalog # 89377-308
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Coiled-coil Domain Containing 107
- Antigen Symbol:CCDC107
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:Coiled-coil domain-containing protein 107.|Ccdc107|CC107_HUMAN
- Cat. No.:89377-308
- Supplier no.:AP10555C
Specifications
About this item
Type: Primary
Antigen: CCDC107
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human