Anti-ZNF211 Rabbit Polyclonal Antibody
89377-290
: Abgent
- Antibody Type:Primary
- Antigen Name:Zinc Finger Protein 211
- Antigen Symbol:ZNF211
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-290
- Supplier no.:AP10546A
Type: Primary
Antigen: ZNF211
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human