Anti-OR52A1 Rabbit Polyclonal Antibody
89377-278
: Abgent
- Antibody Type:Primary
- Antigen Name:Olfactory Receptor, Family 52, Subfamily A Member 1
- Antigen Symbol:OR52A1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-278
- Supplier no.:AP10540B
Type: Primary
Antigen: OR52A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human