Anti-LRRC57 Rabbit Polyclonal Antibody
Catalog # 89377-272
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Leucine Rich Repeat Containing 57
- Antigen Symbol:LRRC57
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-272
- Supplier no.:AP10537B
Specifications
About this item
Type: Primary
Antigen: LRRC57
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human