Anti-DMRT3 Rabbit Polyclonal Antibody
89377-248
: Abgent
- Antibody Type:Primary
- Antigen Name:Doublesex And Mab-3 Related Transcription Factor 3
- Antigen Symbol:DMRT3
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:DMRTA3
- Cat. No.:89377-248
- Supplier no.:AP10526B
Type: Primary
Antigen: DMRT3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse