Anti-IRF2BP2 Rabbit Polyclonal Antibody
Catalog # 89377-244
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:IRF2BP2
- Antigen Symbol:IRF2BP2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-244
- Supplier no.:AP10524C
Specifications
About this item
Type: Primary
Antigen: IRF2BP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human