Anti-ZNF169 Rabbit Polyclonal Antibody
Catalog # 89377-204
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Zinc Finger Protein 169
- Antigen Symbol:ZNF169
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cat. No.:89377-204
- Supplier no.:AP10508A
Specifications
About this item
Type: Primary
Antigen: ZNF169
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human