Anti-ATP6V0C Rabbit Polyclonal Antibody
Catalog # 89377-126
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:ATPase, H+ transporting, lysosomal 16kDa, V0 Subunit c
- Antigen Symbol:ATP6V0C
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Antigen Synonyms:ATP6V0C
- Cat. No.:89377-126
- Supplier no.:AP10470B
Specifications
About this item
Type: Primary
Antigen: ATP6V0C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse