Anti-NRIP3 Rabbit Polyclonal Antibody
100 µg
Catalog # 89376-066
Supplier: Abgent
Supplier #: AP10003B
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Type: Primary
Antigen: NRIP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
Specifications
- Size:100 µg
- Cat. No.:89376-066
- Clonality:Polyclonal
- Reactivity:Human
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:NRIP3
- Host:Rabbit
- Antigen Synonyms:NY-SAR-105|sarcoma antigen NY-SAR-105|C11orf14|nuclear receptor-interacting protein 3
- Antigen Name:Nuclear Receptor-interacting Protein 3
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



