2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec standard
Supplier: BeanTown Chemical
Certificates
About this item
EC No: 244-521-3.
Specifications
- CAS No.:21674-38-4
- MDL No.:MFCD00042439
- Synonym(s):2,4,6-Tris(pentadecafluoroheptyl)-1,3,5-triazine
- Melting Point:26…29 °C
- Formula:C₂₄F₄₅N₃
- Description:2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine