N-Methylmorpholine-N-oxide 50% (w/w) in water
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:7529-22-8
:
Formula:C₅H₁₁NO₂
MDL Number:MFCD00005947
:117.15 g/mol
Boiling Pt:118.5 °C
Melting Pt:–20 °C
Density:1.13 g/mL
EINECS:231-391-8