Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Polyethyleneimine
Polyethyleneimine solution, M.W. ≈ 70,000 30% (w/w) in water, branched
Polyethyleneimine solution, M.W. ≈ 70,000 30% (w/w) in water, branched
Polyethyleneimine solution, M.W. ≈ 70,000 30% (w/w) in water, branched
Supplier:
BeanTown Chemical
Certificates
Expand image
Polyethyleneimine solution, M.W. ≈ 70,000 30% (w/w) in water, branched
Supplier:
BeanTown Chemical
About this item
CAS Number:
9002-98-6
:
Formula:
(CH2CH2NH)n
MDL Number:
MFCD00084427
Related Information
Documentation
SDS
ghs_219710
Documentation