$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Nitric acid
Nitric acid 0.1 N in water volumetric solution
Nitric acid 0.1 N in water volumetric solution
Nitric acid 0.1 N in water volumetric solution
Supplier:
BeanTown Chemical
Certificates
Expand image
Nitric acid 0.1 N in water volumetric solution
Supplier:
BeanTown Chemical
About this item
CAS Number:
7697-37-2
:
Formula:
HNO₃
MDL Number:
MFCD00011349
:
63.01
EINECS:
231-714-2
UN:
3264
ADR:
8,III
Related Information
Documentation
SDS
ghs_218740
Documentation