Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Adenosine ≥99%

Adenosine ≥99%

Adenosine ≥99%
Supplier:  BeanTown Chemical
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CAS: 58-61-7; EC No: 200-389-9; MDL No: MFCD00005752; RTECS: AU7175000 Solid; Molecular Formula: C10H13N5O4; MW: 267.24 Melting Point: 234-236°

  • RTECS: AU7175000
  • Solid
Synonyms:  9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside
  • CAS Number:58-61-7
  • Formula:C₁₀H₁₃N₅O₄
  • MDL Number:MFCD00005752
  • Molecular Weight:267.24 g/mol
  • Melting Pt:234…236 °C
  • EINECS:200-389-9

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    99%
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • HDPE bottle
    10 g
    Log in for availability
    Log in for availability
    Each (10g)
    Each (10g) - $36.70
    $36.70
    Each (10g)
       $36.70

    Supplier Number: 216680-10G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • HDPE bottle
    50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $97.30
    $97.30
    Each (50g)
       $97.30

    Supplier Number: 216680-50G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • HDPE bottle
    250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $412.00
    $412.00
    Each (250g)
       $412.00

    Supplier Number: 216680-250G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.