Silica gel, granules 3-8 mesh desiccant
Supplier: BeanTown Chemical
Certificates
About this item
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:>1600 °C
EINECS:231-545-4
Frequently Bought Together