Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Strontium chloride hexahydrate, Technical Grade

Strontium chloride hexahydrate, Technical Grade

Strontium chloride hexahydrate, Technical Grade
Supplier:  BeanTown Chemical
Certificates
Strontium chloride hexahydrate, Technical Grade

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 10025-70-4; EC No: 233-971-6; MDL No: MFCD00149865 Crystalline; Linear Formula: SrCl2·6H2O ; MW: 266.62 Density (g/mL): 1.93; Refractive Index: 1.650 Hygroscopic

  • Refractive index: 1.650
  • Hygroscopic
  • Crystalline
  • CAS Number:10025-70-4
  • Formula:SrCl₂·6H₂O
  • MDL Number:MFCD00149865
  • Molecular Weight:266.62 g/mol
  • Density:1.93 g/mL
  • EINECS:233-971-6

Documents

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • HDPE bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $71.40
    $71.40
    Each (500g)
       $71.40

    Supplier Number: 213250-500G
  • HDPE bottle
    2 kg
    Log in for availability
    Log in for availability
    Each (2kg)
    Each (2kg) - $166.00
    $166.00
    Each (2kg)
       $166.00

    Supplier Number: 213250-2KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.