Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Polydimethylsiloxane emulsion

Polydimethylsiloxane emulsion
Supplier:  BeanTown Chemical
Certificates
Polydimethylsiloxane emulsion

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL No: MFCD00132673 Liquid

  • Liquid
Synonyms:  Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
  • CAS Number:63148-62-9
  • Formula:(CH₃)₃SiO(Si(CH₃)₂O)n
  • MDL Number:MFCD00084411
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $43.30
    $43.30
    Each (100g)
       $43.30

    Supplier Number: 144165-100G
  • 500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $57.70
    $57.70
    Each (500g)
       $57.70

    Supplier Number: 144165-500G
  • 2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $165.00
    $165.00
    Each (2,500g)
       $165.00

    Supplier Number: 144165-2.5KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.