Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Palladium hydroxide ≥99%

Palladium hydroxide ≥99%

Palladium hydroxide ≥99%
Supplier:  BeanTown Chemical
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 12135-22-7; EC No: 235-219-2; MDL No: MFCD00064599 Powder; Linear Formula: Pd(OH)2; MW: 140.42

  • Powder
  • CAS Number:12135-22-7
  • Formula:H₂O₂Pd
  • MDL Number:MFCD00064599
  • Molecular Weight:140.435 g/mol
  • EINECS:235-219-2

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    99%
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 250 mg
    Log in for availability
    Log in for availability
    Each (250mg)
    Each (250mg) - $65.50
    $65.50
    Each (250mg)
       $65.50

    Supplier Number: 140370-250MG
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $196.00
    $196.00
    Each (1g)
       $196.00

    Supplier Number: 140370-1G
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $808.00
    $808.00
    Each (5g)
       $808.00

    Supplier Number: 140370-5G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.