Potassium Chloride Conductivity Standard, 0.010 M in Water
Supplier: BeanTown Chemical
Certificates
About this item
CAS: 7447-40-7
EC No: 231-211-8
Linear formula: KCl
Form: Liquid
Vapor pressure at 20 °C (68 °F): 23 hPa (17.3 mm Hg)
Density at 20 °C (68 °F): 1 g/cm³ (8.345 lbs./gal)
Solvent content: Water: 99.9%
Solids content: 0.1%
Delivery: Non-hazardous for shipping.
Specifications
- MDL No.:MFCD00011360