Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium stearate

Calcium stearate

Calcium stearate
Supplier:  BeanTown Chemical
Certificates
Calcium stearate
Calcium stearate

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 1592-23-0; EC No: 216-472-8; MDL No: MFCD00036390 Powder; Linear Formula: Ca[O2C(CH2)16CH3]2; Molecular Formula: C26H70CaO4; MW: 607.04

  • Powder
Synonyms:  Stearic acid calcium salt
  • CAS Number:1592-23-0
  • Formula:C₃₆H₇₀CaO₄
  • MDL Number:MFCD00036390
  • Molecular Weight:607.03 g/mol
  • EINECS:216-472-8

Documents

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • HDPE bottle
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $44.60
    $44.60
    Each (1kg)
       $44.60

    Supplier Number: 133410-1KG
  • HDPE bottle
    5 kg
    Log in for availability
    Log in for availability
    Each (5kg)
    Each (5kg) - $151.00
    $151.00
    Each (5kg)
       $151.00

    Supplier Number: 133410-5KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.