Order Entry
Puerto Rico
ContactUsLinkComponent
Zinc selenide, crystals optical disc, polished both sides, Length 25 mm, Width 2 mm
Zinc selenide, crystals optical disc, polished both sides, Length 25 mm, Width 2 mm
Catalog # BT132570-1EACH
Supplier:  BeanTown Chemical
CAS Number:  
Zinc selenide, crystals optical disc, polished both sides, Length 25 mm, Width 2 mm
Catalog # BT132570-1EACH
Supplier:  BeanTown Chemical
Supplier Number:  132570-1EACH
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 each
  • Cat. No.:
    BT132570-1EACH
  • Supplier no.:
    132570-1EACH

Specifications

About this item

 :
 :144.35 g/mol
EINECS:215-259-7
UN:3283
ADR:6.1,III
CAS Number:1315-09-9
Formula:SeZn
MDL Number:MFCD00011296