Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
(2-Chlorophenyl)acetic acid methyl ester ≥98%

(2-Chlorophenyl)acetic acid methyl ester ≥98%

(2-Chlorophenyl)acetic acid methyl ester ≥98%
Supplier:  BeanTown Chemical
Certificates
(2-Chlorophenyl)acetic acid methyl ester ≥98%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 57486-68-7; EC No: 260-764-8; MDL No: MFCD00137445 Liquid; Molecular Formula: C9H9ClO2; MW: 184.62 Boiling Point: 240° Density (g/mL): 1.197

  • Liquid
Synonyms:  (2-Chlorophenyl)acetic acid methyll ester, Methyl 2-chlorophenylacetate, Methyl (2-Chlorophenyl)acetate, Methyl (2-chlorophenyl)acetate
  • CAS Number:57486-68-7
  • Formula:(C₆H₄Cl)CH₂COOCH₃
  • MDL Number:MFCD00137445
  • Molecular Weight:184.62
  • Boiling Pt:240 °C
  • Density:1.197 g/mL
  • EINECS:260-764-8

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    98%
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $133.00
    $133.00
    Each (25g)
       $133.00

    Supplier Number: 130580-25G
  • 100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $377.00
    $377.00
    Each (100g)
       $377.00

    Supplier Number: 130580-100G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.