Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Molecular sieve 3A (0.3 nm, 3 Å)
Molecular sieve 3A (0.3 nm, 3 Å), beads 1-2 mm (0.04-0.08 in)
Molecular sieve 3A (0.3 nm, 3 Å), beads 1-2 mm (0.04-0.08 in)
Molecular sieve 3A (0.3 nm, 3 Å), beads 1-2 mm (0.04-0.08 in)
Supplier:
BeanTown Chemical
Certificates
Expand image
Molecular sieve 3A (0.3 nm, 3 Å), beads 1-2 mm (0.04-0.08 in)
Supplier:
BeanTown Chemical
About this item
CAS Number:
308080-99-1
:
1318-02-1
Formula:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:
MFCD00131613
Related Information
Documentation
SDS
ghs_128945.pdf
Documentation