Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iodine
Iodine 0.1 N in water volumetric solution
Iodine 0.1 N in water volumetric solution
Iodine 0.1 N in water volumetric solution
Supplier:
BeanTown Chemical
Certificates
Expand image
Iodine 0.1 N in water volumetric solution
Supplier:
BeanTown Chemical
Certificates
About this item
CAS Number:
7553-56-2
:
Formula:
I₂
MDL Number:
MFCD00011355
:
253.81 g/mol
EINECS:
231-442-4
UN:
3264
ADR:
8,II
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_128225.pdf
Documentation