Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Potassium citrate monohydrate ≥99%

Potassium citrate monohydrate ≥99%
 
Potassium citrate monohydrate ≥99%
Potassium citrate monohydrate ≥99%

 

 
   
 

CAS: 6100-05-6; EC No: 212-755-5; MDL No: MFCD00150442 Granular; Linear Formula: HOC(COOK)(CH2COOK)2·H2O; Molecular Formula: K3C6H5O7·H2O ; MW: 324.42 Melting Point: 275° (decomposes) Density (g/mL): 1.98 Hygroscopic

 :  Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate
  •  :6100-05-6
  •  :HOC(COOK)(CH₂COOK)₂·H₂O
  •  :MFCD00150442
  •  :324.41 g/mol
  •  :275 °C (dec.)
  •  :1.98 g/ml
  •  :212-755-5
 

 

  • Assay:
    99%
Product Family Options

 

  • Pack type
    Size
     
     
  • HDPE bottle
    500 g
       
       
    Each (500g)
    Each (500g) - $30.20
    $30.20
    Each (500g)
       $30.20

     : 127810-500G
  • HDPE bottle
    2 kg
       
       
    Each (2kg)
    Each (2kg) - $104.00
    $104.00
    Each (2kg)
       $104.00

     : 127810-2KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.