Polystyrene Standard, M.W ≈1,300, MW/MN = 1.06
Catalog # BT127055-1G
Supplier: BeanTown Chemical
CAS Number:
Specifications
- CAS No.:9003-53-6
- Size:1 g
- MDL No.:MFCD00084450
- Synonym(s):PS
- Formula:C₈H₈
- Description:Polystyrene Standard
- Cat. No.:BT127055-1G
- Supplier no.:127055-1G