Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Anionic Standards
Silicate Standards
Silica, Standard Solution, SiO₂ 1000 µg/ml
Silica, Standard Solution, SiO₂ 1000 µg/ml
Silica, Standard Solution, SiO₂ 1000 µg/ml
Supplier:
BeanTown Chemical
Certificates
Expand image
Silica, Standard Solution, SiO₂ 1000 µg/ml
Supplier:
BeanTown Chemical
About this item
Specifications
MDL No.:
MFCD00011232
Solubility:
Fully miscible
Physical Form:
Liquid
Specifications
Related Information
Documentation
SDS
ghs_135425
Documentation