Order Entry
Puerto Rico
ContactUsLinkComponent
Cesium carbonate ≥99% (trace metals basis)
Cesium carbonate ≥99% (trace metals basis)
Catalog # BT123350-50G
Supplier:  BeanTown Chemical
CAS Number:  
Cesium carbonate ≥99% (trace metals basis)
Catalog # BT123350-50G
Supplier:  BeanTown Chemical
Supplier Number:  123350-50G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    HDPE bottle
  • Size:
    50 g
  • Cat. No.:
    BT123350-50G
  • Supplier no.:
    123350-50G

Specifications

Specification Test Results

  • Assay:
    99%

Specification Test Results

About this item

 :
 :325.82 g/mol
Melting Pt:610 °C (dec.)
EINECS:208-591-9
CAS Number:534-17-8
Formula:Cs₂CO₃
MDL Number:MFCD00010957