- Resolution:.001" (.025 mm)
- Description:Digital Calipers
- Cat. No.:100230-754
- Supplier no.:58750
The Digital Callipers display readings in inches or metric at the touch of a button
The Floating zero feature allows scale to read plus or minus a set point without calculations. Stainless steel. Lockable.