Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Apremilast
Apremilast
Apremilast
Apremilast
Supplier:
Avantor
Certificates
Expand image
Apremilast
Supplier:
Avantor
About this item
CAS Number:
608141-41-9
:
Formula:
C₂₂H₂₄N₂O₇S
MDL Number:
MFCD18782607
:
460.5 g/mol
Specification Test Results
CAS:
608141-41-9
Purity:
≥98% by HPLC
Form:
White solid
Specification Test Results
Related Information
Documentation
Manual
2352.pdf
Documentation