Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fenbendazole
Fenbendazole
Fenbendazole
:
Avantor
Fenbendazole
:
Avantor
:
43210-67-9
:
:
C₁₅H₁₃N₃O₂S
:
MFCD00144301
:
299.35 g/mol
CAS:
43210-67-9
Purity:
≥98% by TLC
Form:
White to off-white solid
2157.pdf