Order Entry
Puerto Rico
ContactUsLinkComponent
Gemcitabine hydrochloride
Gemcitabine hydrochloride
Supplier:  Avantor
Certificates
undefined
Gemcitabine hydrochloride
Supplier:  Avantor

Specification Test Results

  • CAS:
    122111-03-9
  • Purity:
    ≥98% by HPLC
  • Form:
    White solid

Specification Test Results

Related Information