Fingolimod hydrochloride
Supplier: Avantor
Certificates
About this item
CAS Number:162359-56-0
:
Formula:C₁₉H₃₄ClNO₂
MDL Number:MFCD00939512
:343.936 g/mol
Specification Test Results
- CAS:162359-56-0
- Purity:≥98% by HPLC
- Form:White solid
Specification Test Results
Frequently Bought Together