Transformer Accessory Kit
Transformer Accessory Kit
Catalog # 470157-638
Supplier: Wards
Supplier #: PH0849AK2
Jump to:
Product Details & Documents
Demonstrate Lenz's Law!
- Excellent accessory to a variable gap magnet
Lenz’s law transformer accessory kit to demonstrate the production of large magnetic fields and lenz’s law damping on pendulums with and without slits.
Specifications
- Description:Transformer Accessory Kit