Carts and Buckets for Cleanroom Use, Texwipe®
Supplier: Texwipe
Certificates
About this item
Stainless steel cart for transporting buckets and bucket systems. Compatible with all stainless steel Texwipe™ buckets.
- Autoclave safe
- Two bucket cleaning system
- Use with cleaning agents, disinfectants, and other solutions in cleanroom environments
- Compatible with all cleanroom string mops and most wringers
This cart has a protective edge to keep your buckets in place and capture spills.
Can add extender to convert into a 3 bucket system (TX7046E).