Order Entry
Puerto Rico
ContactUsLinkComponent
LYFO DISK Bacillus subtilis subsp. subtilis ATCC® 6051™, Microbiologics
LYFO DISK Bacillus subtilis subsp. subtilis ATCC® 6051™, Microbiologics
Catalog # 89502-374
CAS Number:  
LYFO DISK Bacillus subtilis subsp. subtilis ATCC® 6051™, Microbiologics
Catalog # 89502-374
Supplier Number:  0120L
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    LYFO DISK Bacillus subtilis subsp. Subtilis
  • Storage Conditions:
    Easy storage; no dry ice required; reduces shipping costs
  • Traceability:
    Assured Quality - Traceable to reference culture
  • Cat. No.:
    89502-374
  • ATCC no.:
    ATCC® 6051™*
  • Supplier no.:
    0120L

Specifications

About this item

Microorganisms for culture purposes and quality control applications.

  • Easy to use
  • Simply re-hydrate the pellet and inoculate

LYFO DISK® is a lyophilized pellet containing a single strain of a QC microorganism and is the most economical reference stock culture option.

Certifications: LYFO DISK® falls under the category of In-Vitro Diagnostics Medical Devices Directives for CE Mark Conformity. This product is also listed with the FDA.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Packaging: LYFO DISK® is available in a re-sealable vial containing 6 pellets of a single QC microorganism strain.