ContactUsLinkComponent
PowerPac™ HV High-Voltage Power Supply, Bio-Rad
 :  Bio-Rad
PowerPac™ HV High-Voltage Power Supply, Bio-Rad
 :  Bio-Rad
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Catalog No:
  • 100446-702
  • 100446-700
  • Volt-Hours:
  • 99,999 Vh
  • Display:
  • 128×64 pixel, yellow-green backlit graphics LCD
  • Programmable:
  • Stores up to nine basic and nine IEF methods, each with up to nine steps
  • Dimensions:
  • 7.5×34×10 cm (3×13.4×3.9")
  • Input:
  • 90 - 120 or 198 - 264 VAC, 50/60 Hz, autoswitching
  • Operating conditions:
  • 0...40 °C, 0 - 90% humidity in the absence of condensation
  • Output:
  • Four sets in parallel
  • Timer:
  • 1 min–99 hr 59 min, fully adjustable
  • Weight:
  • 6.3 lbs. (2.85 kg)