- Optical property:Optically clear
- Description:Microseal® 'C' Optical Seals, Pack of 100
- Cat. No.:101693-716
- Supplier no.:MSC1001
Microseal® 'C' Optical Seals are optically clear adhesive films designed for real-time PCR and other optical assays.
The seals provide a tight seal on microplates with raised well rims. Microseal® 'C' Optical Seals are free of DNase, RNase, and human DNA contaminants.
The pressure-sensitive adhesive sticks to the plate, not your gloves.