About this item
Individually packaged ReadyAgarose™ Precast Gels are prepared in UV-transparent gel trays designed to fit securely in Bio-Rad’s Mini-Sub® Cell GT and Wide Mini-Sub® Cell GT Cells
Mini ReadyAgarose™ Gels (7.1 x 10 cm) fit securely in any of Bio-Rad's Mini-Sub® Cell GT Systems including Wide Mini ReadySub-Cell™ GT Cells. Choose from 8-, 12-, or 2 x 8-well format, 1% and 3% agarose, TBE buffer with ethidium bromide, and TAE buffer with or without ethidium bromide.
Wide Mini ReadyAgarose™ Gels (15.6 x 10 cm) fit securely in any of Bio-Rad's Wide Mini-Sub® Cell GT Systems including Wide Mini ReadySub-Cell™ GT Cells. Choose from 20-, 32-, 2 x 32-, or 4 x 26-well format, 1% and 3% agarose, TBE buffer and TAE buffer with ethidium bromide, and multichannel pipet compatibility (32-, 2 x 32-, and 4 x 26-well).
ReadyAgarose™ 96 Plus Gels (15 x 10 cm) fit securely in any of Bio-Rad's Wide Mini-Sub® Cell GT Systems including Wide Mini ReadySub-Cell™ GT Cells. These gels resolve nucleic acids from 20 to 20,000 bp, and are designed for high-throughput DNA electrophoretic applications. They have a 4 x 26-well format (96 wells and 8 reference lanes) and are multichannel pipet compatible. Choose from 1% and 3% agarose, and TBE buffer or TAE buffer with ethidium bromide.
ReadyAgarose™ Precast Gels will also fit other manufacturers' electrophoresis cells. The UV-transparent trays have fluorescent well numbers and a ruler printed on them for easy reference, and lock tabs to prevent movement during loading and running. ReadyAgarose™ Gels come in a choice of 27 gel types.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 101643-710
- 101643-684
- 101643-704
- 101643-668
- 101643-674
- 101643-658
- 101643-670
- 101643-700
- 101643-708
- 101643-692
- 101643-680
- 101643-666
- 101643-696
- 101643-662
- 101643-682
- 101643-712
- Gel Tray Dimensions:
- 15 x 10 cm
- 15.6 x 10 cm
- 7.1 x 10 cm
- 7.1 x 10 cm
- 7.1 x 10 cm
- 7.1 x 10 cm
- 7.1 x 10 cm
- 15.6 x 10 cm
- 15 x 10 cm
- 15.6 x 10 cm
- 15.6 x 10 cm
- 7.1 x 10 cm
- 15.6 x 10 cm
- 7.1 x 10 cm
- 15.6 x 10 cm
- 15 x 10 cm