The Wide Mini-Sub® Cell GT Cell is suited for multiple-sample, rapid-screening applications
- QuickSnap electrodes allow easy removal, simplifying cleaning
- Arrow on side of the base indicates run direction, ensuring proper orientation
- Tabs on base allow easy removal of lid, reduce buffer spillage, and prevent incorrect lid positioning
- Accessories include gel casting gates to cast gels right in the cell, or a caster for tape-free casting
- Combs are available to fit every need
- UV-transparent gel trays included
All cells accommodate Bio-Rad's ReadyAgarose™ Precast Gels to save time and allow highly reproducible separations.
The wide platform can accommodate 30 samples per comb. The Wide Mini-Sub® Cell GT Cell is the same width as the Sub-Cell® GT Cell, therefore comb holders, combs, and 15 x 10 cm gel trays are interchangeable with the larger Sub-Cell® GT Cells.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Catalog No:
- 101645-094
- 101645-082
- 100446-682
- 101645-100
- 101645-084
- 101644-988
- Gel Size W×L:
- Wide Mini and 96 Plus Formats
- Wide Mini and 96 Plus Formats
- Wide Mini and 96 Plus Formats
- Wide Mini and 96 Plus Formats
- Wide Mini and 96 Plus Formats
- Wide Mini and 96 Plus Formats
- Buffer Volume:
- ~650 ml
- ~650 ml
- ~650 ml
- ~650 ml
- ~650 ml
- ~650 ml
- Cell Dimensions:
- 17.8 x 25.5 x 6.8
- 17.8 x 25.5 x 6.8
- 17.8 x 25.5 x 6.8
- 17.8 x 25.5 x 6.8
- 17.8 x 25.5 x 6.8
- 17.8 x 25.5 x 6.8
- Migration Speed:
- ~4.5 cm/hr (at 75 V)
- ~4.5 cm/hr (at 75 V)
- ~4.5 cm/hr (at 75 V)
- ~4.5 cm/hr (at 75 V)
- ~4.5 cm/hr (at 75 V)
- ~4.5 cm/hr (at 75 V)
- Sample Throughput:
- 10–60
- 10–60
- 10–60
- 10–60
- 10–60
- 10–60