Human Recombinant GRO-A (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Growth regulated protein alpha (GRO-α), also known as CXCL1, is a chemokine that has mitogenic properties and is a neutrophil chemoattractant. GRO-α is secreted by macrophages, epithelial cells, neutrophils, and melanomas. GRO-α signals through the CXCR2 chemokine receptor and is important during spinal cord formation, inflammation, angiogenesis, tumorigenesis, and wound healing.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P09341
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein Synonyms:CXCL1|mKC|mgSAα|GRO1|NAP-3|rCINC
- Protein/Peptide Name:GRO-A
- Purity:≥95%
- Molecular Weight:7.9 kDa
- Sequence:ASVATELRCQ CLQTLQGIHP KNIQSVNVKS PGPHCAQTEV IATLKNGRKA CLNPASPIVK KIIEKMLNSD KSN
- Endotoxin Level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
Specifications
Frequently Bought Together