Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant BCA-1 (from <i>E. coli</i>)

Human Recombinant BCA-1 (from E. coli)

Human Recombinant BCA-1 (from <i>E. coli</i>)
Certificates
Human Recombinant BCA-1 (from <i>E. coli</i>)

5 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Galectin-1 belongs to the lectin family of carbohydrate binding proteins and binds glycans as both a monomer and a homodimer. Galectin-1 is produced in peripheral lymphoid organs and inflammatory sites. Galectin-1 plays important roles in acute and chronic inflammatory processes, cell growth, cell proliferation, and induces apoptosis of activated T cells. Galectin-1 also modulates cytokine secretion and inhibits pro-inflammatory cytokine production.

  • High quality
  • Low endotoxin
  • Affordable

Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.

Caution: For research use only.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10787-480
  • 10787-478
  • 10787-474
  • 10821-916
  • 10787-476
  • Protein/Peptide Name:
  • BCA-1
  • BCA-1
  • BCA-1
  • BCA-1
  • BCA-1
  • Protein Synonyms:
  • BLC|CXCL13|BLR1 Ligand
  • BLC|CXCL13|BLR1 Ligand
  • BLC|CXCL13|BLR1 Ligand
  • BLC|CXCL13|BLR1 Ligand
  • BLC|CXCL13|BLR1 Ligand
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • Q53X90
  • Q53X90
  • Q53X90
  • Q53X90
  • Q53X90
  • Sequence:
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
  • Protease-free:
  • N
  • N
  • N
  • N
  • N
  • Animal-Free:
  • N
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • N
  • Carrier-Free:
  • Y
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • N
  • Endotoxin Level:
  • Low
  • Low
  • Low
  • Low
  • Low
  • Endotoxin Content:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • UniProtKB:
  • Q53X90
  • Q53X90
  • Q53X90
  • Q53X90
  • Q53X90
  • Biological Activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • Molecular Weight:
  • 10.2 kDa
  • 10.2 kDa
  • 10.2 kDa
  • 10.2 kDa
  • 10.2 kDa
  • Reconstitution Instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage Conditions:
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • −20 °C
  • Shipping Temperature:
  • RT
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Origin
    Availability
    Price
  • Vial
    5 µg
    US
    Log in for availability
    Log in for availability
    Each (5µG)
    Each (5µG) - $92.72
    $92.72
    Each (5µG)
       $92.72
    Minimum order quantity is 1

    Supplier Number: 100-27-5UG
    Specifications:
    More
    Specifications
    Cat. No.
    10787-480
    Protein/Peptide Name
    BCA-1
    Protein Synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological Activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    10.2 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
  • Vial
    20 µg
    US
    Log in for availability
    Log in for availability
    Each (20µG)
    Each (20µG) - $232.03
    $232.03
    Each (20µG)
       $232.03
    Minimum order quantity is 1

    Supplier Number: 100-27-20UG
    Specifications:
    More
    Specifications
    Cat. No.
    10787-478
    Protein/Peptide Name
    BCA-1
    Protein Synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological Activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    10.2 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
  • Vial
    100 µg
    US
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $579.55
    $579.55
    Each (100µG)
       $579.55
    Minimum order quantity is 1

    Supplier Number: 100-27-100UG
    Specifications:
    More
    Specifications
    Cat. No.
    10787-474
    Protein/Peptide Name
    BCA-1
    Protein Synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological Activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    10.2 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
  • Vial
    500 µg
    US
    Log in for availability
    Log in for availability
    Each (500µG)
    Each (500µG) - $2,427.23
    $2,427.23
    Each (500µG)
       $2,427.23
    Minimum order quantity is 1

    Supplier Number: 100-27-500UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10821-916
    Protein/Peptide Name
    BCA-1
    Protein Synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological Activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    10.2 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
  • Vial
    1 mg
    US
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $4,176.49
    $4,176.49
    Each (1mg)
       $4,176.49
    Minimum order quantity is 1

    Supplier Number: 100-27-1MG
    Specifications:
    More
    Specifications
    Cat. No.
    10787-476
    Protein/Peptide Name
    BCA-1
    Protein Synonyms
    BLC|CXCL13|BLR1 Ligand
    Protein/Peptide Type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    Q53X90
    Sequence
    VLEVYYTSLR CRVQESSVFI PRRFIDRIQI LPRGNGCPRK EIIVWKKNKS IVCVDPQAEW IQRMMEVLRK RSSSTLPVPV FKRKIP
    Protease-free
    N
    Animal-Free
    N
    Nuclease-free
    N
    Carrier-Free
    Y
    Endotoxin-free
    N
    Endotoxin Level
    Low
    Endotoxin Content
    Endotoxin LAL (EU/µg) ≤0.1
    UniProtKB
    Q53X90
    Biological Activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular Weight
    10.2 kDa
    Reconstitution Instructions
    Sterile water at 0.1 mg/ml
    Storage Conditions
    −20 °C
    Shipping Temperature
    RT
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.