Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse Recombinant FGF-Basic (from E. coli)
 
undefined
Mouse Recombinant FGF-Basic (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological Activity:
    Bioactivity Assay: 3T3 cell proliferation, ED₅₀ ≤2.5 ng/ml, Bioactivity ≥4.0×10⁵ units/mg
  • Gene ID:
    P15655
  • Reconstitution Instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    N
  • Protein Synonyms:
    Prostatropin|HBGF-2|FGF2
  • Protein/Peptide Name:
    FGF-Basic
  • Purity:
    ≥95%
  • Molecular Weight:
    16.5 kDa
  • Sequence:
    MPALPEDGGA AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • Endotoxin Level:
    Low
  • Formulation:
    10 mM Sodium phosphate, 50 mM Sodium chloride, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping Temperature:
    RT
  • Tested Applications:
    Bioassay

 

Frequently Bought Together