Specifications
- Antibody Type:Tag
- Antigen Symbol:D Tag
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Immunogen:A synthetic peptide (DYKDDDDK) coupled to KLH.
- Cat. No.:10782-442
- Supplier no.:R-1302-100
Specifications
About this item
The D-tag system utilises a short hydrophilic peptide (DYKDDDDK) that is fused to either the N- or C-terminus of the protein of interest. It can be used in conjunction with other tags such as the 6X His tag.
Anti D-tag antibody recognises over-expressed proteins containing the D-tag fused to either amino- or carboxy-termini of targeted proteins in transfected mammalian cells.
Application Information:
Western Blotting (WB), Immunohistochemistry (IH) and Immunoprecipitation (IP). Suggested starting dilution for WB of 1:1000, IS and IP at dilutions of 1:200. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Tag
Antigen: D Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: