- Antibody Type:Primary
- Antigen Name:Parathyroid Hormone 2 Receptor
- Antigen Symbol:PTH2R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 µL
- Epitope:481-501 aa
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:Pthr2|Parathyroid hormone receptor|PTH2 receptor
- Immunogen:A synthetic peptide (RQIDSHVTLPGYVWSSSEQDC) of rat Parathyroid hormone receptor protein (aa: 481-501) conjugated to KLH has been used as the immunogen.
- Cat. No.:10782-428
- Supplier no.:R-120-100
FUNCTION: This is a specific receptor for parathyroid hormone. The activity of this receptor is mediated by g proteins which activate adenylyl cyclase. PTHR2 may be responsible for PTH effects in a number of physiological systems. It may play a significant role in pancreatic function. PTHR2 presence in neurons indicates that it may function as a neurotransmitter receptor. SUBUNIT: Binds to TIPF39/TIP39. SUBCELLULAR LOCATION: Membrane; multi-pass membrane protein. TISSUE SPECIFICITY: Abundantly expressed in brain, arterial and cardiac endothelium. Found as well in sperm, in the head of the epididymis. Lower expression is found in vascular smooth muscle, exocrine pancreas, testis and placenta. SIMILARITY: Belongs to the G-protein coupled receptor 2 family.
Specificity has been shown by western blot. A band of 63 kDa corresponding to the theoretical molecular weight of Parathyroid hormone receptor is easily detectable by WB. Absorption of the antiserum with the immunising peptide abolished the binding of the antibody to the target hence detectability of the band.
Application Information:
WB. A dilution of 1:500 to 1:1000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: PTH2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 481-501 aa
Host: Rabbit
Isotype: IgG
Reactivity: Rat