Anti-ATG12 Rabbit Polyclonal Antibody
Catalog # 10782-414
Supplier: Biosensis
Specifications
- Antibody Type:Primary
- Antigen Name:Autophagy Related 12
- Antigen Symbol:ATG12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µL
- Epitope:N-Terminal of Human ATG12
- Cross Adsorption:No
- Format:Lyophilized
- Antigen Synonyms:FBR93|HAPG12|APG12L|APG12
- Immunogen:A synthetic peptide corresponding to the N-terminal of human ATG12 protein has been used as the immunogen. No immunogenic carrier protein was conjugated to the immunogen. Instead, Adjukine B (see the Adjuvants in biosensis' product list) has been used to orchestrate/boost the immune response.
- Cat. No.:10782-414
- Supplier no.:R-112-100
Specifications
About this item
FUNCTION: Required for autophagy. SUBUNIT: Conjugated to ATG5. SUBCELLULAR LOCATION: Cytoplasm. ALTERNATIVE PRODUCTS: 2 named isoforms produced by alternative splicing. TISSUE SPECIFICITY: Ubiquitous. SIMILARITY: Belongs to the ATG12 family.
IHC and WB confirmed the specificity for ATG12.
Application Information:
IHC, immunofluorescence, WB. A dilution of 1:200 to 1:1000 dilution is recommended for these applications. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ATG12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal of Human ATG12
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat